missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444680050
Specifications
3-Chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester | |
5g | |
96% min. (GC) | |
2-chloro-6-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-chloro-6-methoxy-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester, 3-chloro-4-hydroxy-5-methoxyphenylboronic acid, pinacol ester, 6-chloro-2-methoxy-4-4,4,5,5-tetra methyl-1,3,2-dioxaborolan-2-yl phenol, phenol, 2-chloro-6-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)Cl)O)OC | |
284.55 | |
284.55 |
156°C to 159°C | |
1003298-84-7 | |
C13H18BClO4 | |
XBEJPOZGAPQFPL-UHFFFAOYSA-N | |
2-chloro-6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol | |
17750230 | |
97% |
Chemical Identifiers
1003298-84-7 | |
284.55 | |
2-chloro-6-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 2-chloro-6-methoxy-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 3-chloro-4-hydroxy-5-methoxyphenylboronic acid pinacol ester, 3-chloro-4-hydroxy-5-methoxyphenylboronic acid, pinacol ester, 6-chloro-2-methoxy-4-4,4,5,5-tetra methyl-1,3,2-dioxaborolan-2-yl phenol, phenol, 2-chloro-6-methoxy-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
2-chloro-6-methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol |
C13H18BClO4 | |
XBEJPOZGAPQFPL-UHFFFAOYSA-N | |
17750230 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C(=C2)Cl)O)OC |
Safety and Handling
GHS Signal Word: Warning