Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-hydroxy-5-methoxybenzyl alcohol, 98%

$252.45 - $252.45

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-hydroxy-5-methoxybenzyl alcohol
CAS 20624-92-4
Molecular Formula C8H9ClO3
Formula Weight 188.62
MDL Number MFCD00016870
Synonym 5-Chlorovanillyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2510814

  SDS
alfa aesar™
B2510814
25g Each for $252.45
Description & Specifications

Specifications

Formula Weight 188.62
Chemical Name or Material 3-Chloro-4-hydroxy-5-methoxybenzyl alcohol
Assay 98%
Molecular Formula C8H9ClO3
MDL Number MFCD00016870
CAS 20624-92-4
Synonym 5-Chlorovanillyl alcohol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 115° to 118°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.