Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-formylbenzeneboronic acid, 95%

$221.50 - $498.74

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-formylbenzeneboronic acid
CAS 1072952-53-4
Molecular Formula C7H6BClO3
Formula Weight 184.39
MDL Number MFCD08274473
Synonym 4-Borono-2-chlorobenzaldehyde; 3-Chloro-4-formylphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2772503

  SDS
alfa aesar™
H2772503
1g Each for $498.74

AAH27725MD

  SDS
alfa aesar™
H27725MD
250mg Each for $221.50
Description & Specifications

Specifications

Formula Weight 184.39
Chemical Name or Material 3-Chloro-4-formylbenzeneboronic acid
Health Hazard 1 Warning
Assay 95%
MDL Number MFCD08274473
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H6BClO3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 1072952-53-4
Quantity 1g
Melting Point 210° to 214°C
Synonym 4-Borono-2-chlorobenzaldehyde; 3-Chloro-4-formylphenylboronic acid

Applications
Used in suzuki cross-coupling.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.