missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-fluorophenylhydrazine hydrochloride, 98%

Manufacturer:  Alfa Aesar™ A1017522

 View more versions of this product

Catalog No. AAA1017522



Specifications

Specifications

3-Chloro-4-fluorophenylhydrazine hydrochloride
100g
C6H7Cl2FN2
3-chloro-4-fluorophenylhydrazine hydrochloride, 3-chloro-4-fluorophenyl hydrazine hydrochloride, 1-3-chloro-4-fluorophenyl hydrazine hydrochloride, hydrazine, 3-chloro-4-fluorophenyl-, monohydrochloride, pubchem3507, acmc-1c5q6, labotest-bb lt01147590, ksc494o5j, 3-chloro-4-fluorophenylhydrazinehcl, 3-chloro-4-fluorophenylhydrazine hcl
C1=CC(=C(C=C1NN)Cl)F.Cl
197.034
197.04
~210°C (decomposition)
175135-74-7
MFCD00052267
DWCDDXINAZHRLK-UHFFFAOYSA-N
(3-chloro-4-fluorophenyl)hydrazine;hydrochloride
44181777
98%

Chemical Identifiers

175135-74-7
197.034
DWCDDXINAZHRLK-UHFFFAOYSA-N
44181777
C1=CC(=C(C=C1NN)Cl)F.Cl
C6H7Cl2FN2
MFCD00052267
3-chloro-4-fluorophenylhydrazine hydrochloride, 3-chloro-4-fluorophenyl hydrazine hydrochloride, 1-3-chloro-4-fluorophenyl hydrazine hydrochloride, hydrazine, 3-chloro-4-fluorophenyl-, monohydrochloride, pubchem3507, acmc-1c5q6, labotest-bb lt01147590, ksc494o5j, 3-chloro-4-fluorophenylhydrazinehcl, 3-chloro-4-fluorophenylhydrazine hcl
(3-chloro-4-fluorophenyl)hydrazine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

SDS Documents