missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-4-fluorophenylboronic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 144432-85-9
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H5BClFO2
Formula Weight 174.37
MDL Number MFCD00051800
Chemical Name or Material 3-Chloro-4-fluorophenylboronic acid
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC367680050 SDS Thermo Fisher Scientific
AC367680050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

144432-85-9
100.0
C6H5BClFO2
MFCD00051800
≥96.0%
96.0
97%
174.37
3-Chloro-4-fluorophenylboronic acid

Specifications

Authentic
5g
96.0
100.0
97%
174.37
3-Chloro-4-fluorophenylboronic acid
Off-white to faintly orange
144432-85-9
Glass bottle
Powder
C6H5BClFO2
MFCD00051800
≥96.0%