missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-fluorophenylacetic acid, 98%

$60.66 - $60.66

Chemical Identifiers

CAS 705-79-3
Molecular Formula C8H6ClFO2
Formula Weight 188.59
MDL Number MFCD01631555
Chemical Name or Material 3-Chloro-4-fluorophenylacetic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH26114MD SDS Alfa Aesar™
H26114MD
250mg Each for $60.66
AAH2611403 SDS Alfa Aesar™
H2611403
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

705-79-3
188.59
3-Chloro-4-fluorophenylacetic acid
C8H6ClFO2
MFCD01631555

Specifications

55° to 58°C
705-79-3
188.59
3-Chloro-4-fluorophenylacetic acid
250mg
C8H6ClFO2
MFCD01631555
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning