missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-chloro-4-fluorophenyl isocyanate, 98%, ACROS Organics™

$27.79 - $27.79

Chemical Identifiers

CAS 50529-33-4
Molecular Formula C7H3ClFNO
Molecular Weight (g/mol) 171.56
MDL Number MFCD00037037
InChI Key XVIPJBUXMFLHSI-UHFFFAOYSA-N
Synonym 3-chloro-4-fluoro phenyl isocyanate, 3-chloro-4-fluoro-phenylisocyanate, 3-chloro-4-fluorobenzenisocyanate, 3-chloro-4-fluorophenyl isocyanate, 3-chloro-4-fluorophenyl-isocyanate, 3-chloro-4-fluorophenylisocyanate, acmc-20an6r, benzene, 2-chloro-1-fluoro-4-isocyanato, pubchem5024
PubChem CID 2734632
IUPAC Name 2-chloro-1-fluoro-4-isocyanatobenzene
SMILES C1=CC(=C(C=C1N=C=O)Cl)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC366790010
SDS View Documents
Thermo Fisher Scientific
AC366790010
1g Glass Bottle
Each for $27.79
 
Description

Description

Specifications

Chemical Identifiers

50529-33-4
171.56
XVIPJBUXMFLHSI-UHFFFAOYSA-N
2734632
C1=CC(=C(C=C1N=C=O)Cl)F
C7H3ClFNO
MFCD00037037
3-chloro-4-fluoro phenyl isocyanate, 3-chloro-4-fluoro-phenylisocyanate, 3-chloro-4-fluorobenzenisocyanate, 3-chloro-4-fluorophenyl isocyanate, 3-chloro-4-fluorophenyl-isocyanate, 3-chloro-4-fluorophenylisocyanate, acmc-20an6r, benzene, 2-chloro-1-fluoro-4-isocyanato, pubchem5024
2-chloro-1-fluoro-4-isocyanatobenzene

Specifications

1.3770g/mL
Authentic
1.5395 to 1.5415
44°C (17.0mmHg)
1g
97.5
97.5% min. (GC)
Cl(F)C6H3NCO
3-chloro-4-fluoro phenyl isocyanate, 3-chloro-4-fluoro-phenylisocyanate, 3-chloro-4-fluorobenzenisocyanate, 3-chloro-4-fluorophenyl isocyanate, 3-chloro-4-fluorophenyl-isocyanate, 3-chloro-4-fluorophenylisocyanate, acmc-20an6r, benzene, 2-chloro-1-fluoro-4-isocyanato, pubchem5024
C1=CC(=C(C=C1N=C=O)Cl)F
171.56
171.56
98%
87°C
Glass Bottle
1.377
Colorless to Yellow
50529-33-4
100.0
C7H3ClFNO
MFCD00037037
XVIPJBUXMFLHSI-UHFFFAOYSA-N
2-chloro-1-fluoro-4-isocyanatobenzene
2734632
Liquid
3-Chloro-4-fluorophenyl isocyanate, 98%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C

SDS