Please login to your online account to display your discounted pricing

3-chloro-4-fluorophenyl isocyanate, 98%, ACROS Organics™


Quantity 1g
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  AC366790010

 View more versions of this product

Catalog No. AC366790010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 171.56
Quantity 1g
Color Clear colorless to slightly yellow
Density 1.3770g/mL
CAS Max % 100.0
Chemical Name or Material 3-Chloro-4-fluorophenyl isocyanate
Molecular Formula C7H3ClFNO
MDL Number MFCD00037037
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Infrared Spectrum Authentic
Name Note 98%
Packaging Glass Bottle
Recommended Storage Refrigerator +4°C
Refractive Index 1.5395 to 1.5415 (20°C, 589nm)
Assay Percent Range 98%
Linear Formula Cl(F)C6H3NCO
CAS 50529-33-4

Specifications

Chemical Name or Material 3-Chloro-4-fluorophenyl isocyanate
Name Note 98%
CAS 50529-33-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H3ClFNO
Linear Formula Cl(F)C6H3NCO
Formula Weight 171.56
MDL Number MFCD00037037
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

Refrigerator +4°C

SDS & Safety Documents