missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-4-fluorophenol, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 277200250

 View more versions of this product

Catalog No. AC277200250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Chloro-4-fluorophenol
Authentic
104°C (11.0mmHg)
39°C to 44°C
99%
98.5
99%
ClC6H3FOH
3-chloro-4-fluoro-phenol, 3-chloro-4-fluorophe, 3-chloro-4-fluorophenol, 3-chloro4-fluorophenol, 4-fluoro-3-chlorophenol, acmc-209goi, ksc204s3j, phenol, 3-chloro-4-fluoro, pubchem1492, pubchem2856
ZQXLIXHVJVAPLW-UHFFFAOYSA-N
3-chloro-4-fluorophenol
75790
Crystalline Solid
108°C
Glass bottle
Beige to Brown
25g
2613-23-2
100.0
C6H4ClFO
MFCD00002257
Solubility in water: slightly soluble. Other solubilities: soluble in most common organic solvents
C1=CC(=C(C=C1O)Cl)F
146.55
146.55
99%

Chemical Identifiers

2613-23-2
146.55
ZQXLIXHVJVAPLW-UHFFFAOYSA-N
75790
C1=CC(=C(C=C1O)Cl)F
C6H4ClFO
MFCD00002257
3-chloro-4-fluoro-phenol, 3-chloro-4-fluorophe, 3-chloro-4-fluorophenol, 3-chloro4-fluorophenol, 4-fluoro-3-chlorophenol, acmc-209goi, ksc204s3j, phenol, 3-chloro-4-fluoro, pubchem1492, pubchem2856
3-chloro-4-fluorophenol
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 220-042-5

TSCA : TSCA

SDS Documents