Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-fluorobenzyl bromide, 97%

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-fluorobenzyl bromide
CAS 192702-01-5
Molecular Formula C7H5BrClF
Formula Weight 223.48
MDL Number MFCD01631551
UN Number UN3265
Beilstein 2435145
Synonym alpha-Bromo-3-chloro-4-fluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1914603

  SDS
alfa aesar™
L1914603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL19146MD

  SDS
alfa aesar™
L19146MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 223.48
Assay 97%
Chemical Name or Material 3-Chloro-4-fluorobenzyl bromide
CAS 192702-01-5
Molecular Formula C7H5BrClF
Health Hazard 1 Danger
Synonym alpha-Bromo-3-chloro-4-fluorotoluene
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
MDL Number MFCD01631551
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 1g
Boiling Point 63° to 65°C (0.6mmHg)
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Beilstein 2435145
UN Number UN3265
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.