Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-fluorobenzoic acid, 98%

$39.78 - $534.91

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-fluorobenzoic acid
CAS 403-16-7
Molecular Formula C7H4ClFO2
Formula Weight 174.56
MDL Number MFCD00042477
Beilstein 2574799
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1281403

  SDS
alfa aesar™
A1281403
1g Each for $39.78

AAA1281406

  SDS
alfa aesar™
A1281406
5g Each for $138.01

AAA1281414

  SDS
alfa aesar™
A1281414
25g Each for $534.91
Description & Specifications

Specifications

Formula Weight 174.56
Molecular Formula C7H4ClFO2
Quantity 1g
CAS 403-16-7
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 133° to 136°C
MDL Number MFCD00042477
Chemical Name or Material 3-Chloro-4-fluorobenzoic acid
Assay 98%
Beilstein 2574799
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.