Please login to your online account to display your discounted pricing

3-Chloro-4-fluorobenzoic acid, 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC308970010

 View more versions of this product

Catalog No. AC308970010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystals or Needles
Formula Weight 174.56
Quantity 1g
Color Off-white to beige
TSCA TSCA
Melting Point 130° to 137°C
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Name Note 95%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Chloro-4-fluorobenzoic acid
Linear Formula ClC6H3(F)CO2H
MDL Number MFCD00042477
Assay Percent Range 95%
Molecular Formula C7 H4 Cl F O2
CAS 403-16-7
Packaging Glass bottle

Specifications

Chemical Name or Material 3-Chloro-4-fluorobenzoic acid
Name Note 95%
CAS 403-16-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7 H4 Cl F O2
Linear Formula ClC6H3(F)CO2H
Formula Weight 174.56
MDL Number MFCD00042477
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents