Please login to your online account to display your discounted pricing

3-Chloro-4-fluorobenzoic acid, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-fluorobenzoic acid
Name Note 95%
CAS 403-16-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C7 H4 Cl F O2
Linear Formula ClC6H3(F)CO2H
Formula Weight 174.56
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC308970010

  SDS
thermo fisher scientific
AC308970010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystals or Needles
Formula Weight 174.56
Quantity 1g
Color Off-white to beige
TSCA TSCA
Melting Point 130° to 137°C
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Name Note 95%
Linear Formula ClC6H3(F)CO2H
MDL Number MFCD00042477
Chemical Name or Material 3-Chloro-4-fluorobenzoic acid
Assay Percent Range 95%
Molecular Formula C7 H4 Cl F O2
CAS 403-16-7
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.