Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-fluorobenzenesulfonamide, 97%

$119.30 - $355.57

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-fluorobenzenesulfonamide
CAS 146533-46-2
Molecular Formula C6H5ClFNO2S
Formula Weight 209.63
MDL Number MFCD00130211
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2640606

  SDS
alfa aesar™
H2640606
5g Each for $355.57

AAH2640603

  SDS
alfa aesar™
H2640603
1g Each for $119.30
Description & Specifications

Specifications

Formula Weight 209.63
Chemical Name or Material 3-Chloro-4-fluorobenzenesulfonamide
Assay 97%
Molecular Formula C6H5ClFNO2S
MDL Number MFCD00130211
CAS 146533-46-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point 107° to 110°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.