Please login to your online account to display your discounted pricing

3-Chloro-4-fluorobenzene-1-thiol, Tech., Maybridge

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-fluorobenzene-1-thiol
Assay Tech.
Molecular Formula C6H4ClFS
Formula Weight 162.61
MDL Number MFCD00052938
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00315DA

  SDS
fisher scientific company
TL00315DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.61
Purity Grade Technical Grade
Quantity 1g
Assay Tech.
Chemical Name or Material 3-Chloro-4-fluorobenzene-1-thiol
Packaging Amber Glass Bottle
CAS 60811-23-6
MDL Number MFCD00052938
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C6H4ClFS
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.