missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-fluorobenzamide, 97%

$78.04 - $281.69

Chemical Identifiers

CAS 701-43-9
Molecular Formula C7H5ClFNO
Formula Weight 173.58
MDL Number MFCD00153092
Chemical Name or Material 3-Chloro-4-fluorobenzamide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1439203 SDS Alfa Aesar™
A1439203
1g Each for $78.04
AAA1439206 SDS Alfa Aesar™
A1439206
5g Each for $281.69
Specifications Description & Specifications

Chemical Identifiers

701-43-9
173.58
3-Chloro-4-fluorobenzamide
C7H5ClFNO
MFCD00153092

Specifications

145° to 147°C
701-43-9
173.58
3-Chloro-4-fluorobenzamide
1g
C7H5ClFNO
MFCD00153092
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning