Please login to your online account to display your discounted pricing

3-chloro-4-fluorobenzaldehyde, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-chloro-4-fluorobenzaldehyde
CAS 34328-61-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4ClFO
Linear Formula ClC6H3(F)CHO
Formula Weight 158.56
MDL Number MFCD00011735
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC311220050

  SDS
thermo fisher scientific
AC311220050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC311220010

  SDS
thermo fisher scientific
311220010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid After Melting
Formula Weight 158.56
Quantity 5g
Color Clear light yellow
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-chloro-4-fluorobenzaldehyde
Assay Percent Range 97%
Molecular Formula C7H4ClFO
Linear Formula ClC6H3(F)CHO
CAS 34328-61-5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
MDL Number MFCD00011735
Packaging Glass bottle
Refractive Index 1.5440 to 1.5460 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.