Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-fluorobenzaldehyde, 96%

$55.76 - $185.34

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-fluorobenzaldehyde
CAS 34328-61-5
Molecular Formula C7H4ClFO
Formula Weight 158.56
MDL Number MFCD00011735
Beilstein 1857885
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1992303

  SDS
alfa aesar™
A1992303
1g Each for $55.76

AAA1992306

  SDS
alfa aesar™
A1992306
5g Each for $185.34
Description & Specifications

Specifications

Formula Weight 158.56
Assay 96%
Molecular Formula C7H4ClFO
MDL Number MFCD00011735
CAS 34328-61-5
Beilstein 1857885
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Melting Point 25° to 28°C
Quantity 1g
Chemical Name or Material 3-Chloro-4-fluorobenzaldehyde
Boiling Point 66° to 72°C (2mmHg)
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.