missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-fluoroanisole, 97%

$37.09 - $478.69

Chemical Identifiers

CAS 202925-07-3
Molecular Formula C7H6ClFO
Formula Weight 160.58
MDL Number MFCD00070778
Chemical Name or Material 3-Chloro-4-fluoroanisole
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2242503 SDS Alfa Aesar™
B2242503
1g Each for $37.09
AAB2242506 SDS Alfa Aesar™
B2242506
5g Each for $124.61
AAB2242514 SDS Alfa Aesar™
B2242514
25g Each for $478.69
Specifications Description & Specifications

Chemical Identifiers

202925-07-3
160.58
3-Chloro-4-fluoroanisole
C7H6ClFO
MFCD00070778

Specifications

79°C (175°F)
1g
C7H6ClFO
MFCD00070778
97%
189°C
202925-07-3
160.58
3-Chloro-4-fluoroanisole

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning