Learn More
3-Chloro-4-fluoroaniline, 98%, Thermo Scientific Chemicals
$274.29 - $274.29
Chemical Identifiers
CAS | 367-21-5 |
---|---|
Molecular Formula | C6H5ClFN |
Molecular Weight (g/mol) | 145.56 |
MDL Number | MFCD00007767 |
InChI Key | YSEMCVGMNUUNRK-UHFFFAOYSA-N |
Synonym | benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin |
PubChem CID | 9708 |
IUPAC Name | 3-chloro-4-fluoroaniline |
SMILES | NC1=CC=C(F)C(Cl)=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC211531000
|
Thermo Scientific Chemicals
211531000 |
100 g | Glass bottle |
Each for $274.29
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
367-21-5 | |
145.56 | |
YSEMCVGMNUUNRK-UHFFFAOYSA-N | |
9708 | |
NC1=CC=C(F)C(Cl)=C1 |
C6H5ClFN | |
MFCD00007767 | |
benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin | |
3-chloro-4-fluoroaniline |
Specifications
42°C to 47°C | |
Beige to Brown | |
100.0 | |
100 g | |
Glass bottle | |
98% | |
ClC6H3(F)NH2 | |
12, I, 309 | |
Solubility in water: 10g/L water 520°C). Other solubilities: soluble in organic solvents | |
NC1=CC=C(F)C(Cl)=C1 | |
145.56 | |
145.56 | |
3-Chloro-4-fluoroaniline, 98% |
367-21-5 | |
97.5 | |
230°C | |
Authentic | |
Crystalline Solid | |
C6H5ClFN | |
MFCD00007767 | |
benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin | |
YSEMCVGMNUUNRK-UHFFFAOYSA-N | |
3-chloro-4-fluoroaniline | |
9708 | |
98% |
Safety and Handling
GHS H Statement
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
EINECSNumber : 206-682-8
TSCA : TSCA
Recommended Storage : Product darkens on prolonged storage
RUO – Research Use Only