missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-fluoroaniline, 98+%

Manufacturer:  Alfa Aesar™ A1185814

 View more versions of this product

Catalog No. AAA1185814



Specifications

Specifications

3-Chloro-4-fluoroaniline
Characteristic
44°C to 47°C
367-21-5
MFCD00007767
1562786
YSEMCVGMNUUNRK-UHFFFAOYSA-N
3-chloro-4-fluoroaniline
9708
≥98%
149°C (300°F)
227°C to 230°C
25g
C6H5ClFN
UN2811
benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin
C1=CC(=C(C=C1N)Cl)F
145.561
145.56

Chemical Identifiers

367-21-5
145.561
YSEMCVGMNUUNRK-UHFFFAOYSA-N
9708
C1=CC(=C(C=C1N)Cl)F
C6H5ClFN
MFCD00007767
benzenamine, 3-chloro-4-fluoro, 3-chloro-4-fluorobenzenamine, 3-chloro-4-fluoro-phenylamine, aniline, 3-chloro-4-fluoro, 3-chloro-4-fluoro aniline, unii-7l63cc70uq, 3-chloro-4-fluorophenylamine, 4-fluoro-3-chloroaniline, 3-chloro-4-fluoro-aniline, 3-chlor-4-fluoranilin
3-chloro-4-fluoroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents