Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Chloro-4'-fluoroacetophenone, 97%

$111.59 - $1,329.93

Chemical Identifiers

Chemical Name or Material 3'-Chloro-4'-fluoroacetophenone
CAS 2923-66-2
Molecular Formula C8H6ClFO
Formula Weight 172.59
MDL Number MFCD00042203
Beilstein 3117772
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1515806

 
alfa aesar™
A1515806
5g Each for $111.59

AAA1515814

 
alfa aesar™
A1515814
25g Each for $406.36

AAA1515822

 
alfa aesar™
A1515822
100g Each for $1,329.93
Description & Specifications

Specifications

Formula Weight 172.59
Chemical Name or Material 3'-Chloro-4'-fluoroacetophenone
Assay 97%
Molecular Formula C8H6ClFO
MDL Number MFCD00042203
CAS 2923-66-2
Beilstein 3117772
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 126° to 127°C (15mmHg)
Melting Point 36° to 42°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.