missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Chloro-4'-fluoroacetophenone, 97%

Manufacturer:  Alfa Aesar™ A1515814

 View more versions of this product

Catalog No. AAA1515814



Specifications

Specifications

3'-Chloro-4'-fluoroacetophenone
36°C to 42°C
2923-66-2
MFCD00042203
3'-chloro-4'-fluoroacetophenone, 3-chloro-4-fluoroacetophenone, 1-3-chloro-4-fluorophenyl ethanone, 1-3-chloro-4-fluorophenyl ethan-1-one, ethanone, 1-3-chloro-4-fluorophenyl, 1-3-chloro-4-fluoro-phenyl-ethanone, 1-acetyl-3-chloro-4-fluorobenzene, pubchem4202, ksc699k7b, 3-chloro-4-fluoro acetophenone
CC(=O)C1=CC(=C(C=C1)F)Cl
172.583
172.59
126°C to 127°C (15mmHg)
25g
C8H6ClFO
3117772
PCJPESKRPOTNGU-UHFFFAOYSA-N
1-(3-chloro-4-fluorophenyl)ethanone
520422
97%

Chemical Identifiers

2923-66-2
172.583
PCJPESKRPOTNGU-UHFFFAOYSA-N
520422
CC(=O)C1=CC(=C(C=C1)F)Cl
C8H6ClFO
MFCD00042203
3'-chloro-4'-fluoroacetophenone, 3-chloro-4-fluoroacetophenone, 1-3-chloro-4-fluorophenyl ethanone, 1-3-chloro-4-fluorophenyl ethan-1-one, ethanone, 1-3-chloro-4-fluorophenyl, 1-3-chloro-4-fluoro-phenyl-ethanone, 1-acetyl-3-chloro-4-fluorobenzene, pubchem4202, ksc699k7b, 3-chloro-4-fluoro acetophenone
1-(3-chloro-4-fluorophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents