missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-(ethylcarbamoyl)benzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5316203

 View more versions of this product

Catalog No. AAH5316203



Specifications

Specifications

3-Chloro-4-(ethylcarbamoyl)benzeneboronic acid
1g
C9H11BClNO3
3-chloro-4-ethylcarbamoyl phenyl boronic acid, 3-chloro-4-n-ethylcarbamoyl phenylboronic acid, 3-chloro-4-n-ethylcarbamoyl benzeneboronic acid, 3-chloro-4-ethylcarbamoyl phenylboronic acid, 3-chloro-4-ethylcarbamoyl phenyl boronic acid, boronic acid,b-3-chloro-4-ethylamino carbonyl phenyl, 3-chloro-4-n-ethyl carbamoyl phenylboronic acid, 3-chloranyl-4-ethylcarbamoyl phenyl boronic acid
B(C1=CC(=C(C=C1)C(=O)NCC)Cl)(O)O
227.451
227.45
142°C to 146°C
850589-40-1
MFCD07363767
HJDYETAQUCUPFP-UHFFFAOYSA-N
[3-chloro-4-(ethylcarbamoyl)phenyl]boronic acid
44119416
97%

Chemical Identifiers

850589-40-1
227.451
HJDYETAQUCUPFP-UHFFFAOYSA-N
44119416
B(C1=CC(=C(C=C1)C(=O)NCC)Cl)(O)O
C9H11BClNO3
MFCD07363767
3-chloro-4-ethylcarbamoyl phenyl boronic acid, 3-chloro-4-n-ethylcarbamoyl phenylboronic acid, 3-chloro-4-n-ethylcarbamoyl benzeneboronic acid, 3-chloro-4-ethylcarbamoyl phenylboronic acid, 3-chloro-4-ethylcarbamoyl phenyl boronic acid, boronic acid,b-3-chloro-4-ethylamino carbonyl phenyl, 3-chloro-4-n-ethyl carbamoyl phenylboronic acid, 3-chloranyl-4-ethylcarbamoyl phenyl boronic acid
[3-chloro-4-(ethylcarbamoyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents