Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-ethoxy-5-fluorophenol, 97%

$306.31 - $306.31

Chemical Identifiers

Chemical Name or Material 3-Chloro-4-ethoxy-5-fluorophenol
Molecular Formula C8H8ClFO2
Formula Weight 190.6
MDL Number MFCD09832339
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3265303

 
alfa aesar™
H3265303
1g Each for $306.31
Description & Specifications

Specifications

Formula Weight 190.6
Chemical Name or Material 3-Chloro-4-ethoxy-5-fluorophenol
Assay 97%
Molecular Formula C8H8ClFO2
MDL Number MFCD09832339
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 86° to 90°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.