missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid, 98%

Manufacturer:  Alfa Aesar™ H5331503

 View more versions of this product

Catalog No. AAH5331503



Specifications

Specifications

3-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid
1g
C11H13BClNO4
3-chloro-4-morpholine-4-carbonyl phenyl boronic acid, 3-chloro-4-morpholine-4-carbonyl phenylboronic acid, 3-chloro-4-morpholine-4-carbonyl benzeneboronic acid, 3-chloro-4-morpholinocarbonyl phenylboronic acid, 3-chloro-4-morpholine-4-carbonyl phenyl boronic acid, 4-4-borono-2-chlorobenzoyl morpholine, n-morpholinyl 2-chloro-4-boronobenzamide, 3-chloro-4-morpholine-4-carbonyl phenyl boronicacid, 4-borono-2-chlorophenyl morpholin-4-yl methanone, 3-chloro-4-4-morpholinylcarbonyl benzeneboronic acid
B(C1=CC(=C(C=C1)C(=O)N2CCOCC2)Cl)(O)O
269.488
253.49
182°C to 186°C
850589-49-0
MFCD07363774
HPRBSCDTOODIQX-UHFFFAOYSA-N
[3-chloro-4-(morpholine-4-carbonyl)phenyl]boronic acid
44119419
98%

Chemical Identifiers

850589-49-0
269.488
HPRBSCDTOODIQX-UHFFFAOYSA-N
44119419
B(C1=CC(=C(C=C1)C(=O)N2CCOCC2)Cl)(O)O
C11H13BClNO4
MFCD07363774
3-chloro-4-morpholine-4-carbonyl phenyl boronic acid, 3-chloro-4-morpholine-4-carbonyl phenylboronic acid, 3-chloro-4-morpholine-4-carbonyl benzeneboronic acid, 3-chloro-4-morpholinocarbonyl phenylboronic acid, 3-chloro-4-morpholine-4-carbonyl phenyl boronic acid, 4-4-borono-2-chlorobenzoyl morpholine, n-morpholinyl 2-chloro-4-boronobenzamide, 3-chloro-4-morpholine-4-carbonyl phenyl boronicacid, 4-borono-2-chlorophenyl morpholin-4-yl methanone, 3-chloro-4-4-morpholinylcarbonyl benzeneboronic acid
[3-chloro-4-(morpholine-4-carbonyl)phenyl]boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents