Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-nitropyridine, 97%

$238.59 - $938.96

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-nitropyridine
CAS 54231-32-2
Molecular Formula C5H3ClN2O2
Formula Weight 158.54
MDL Number MFCD00490110
Beilstein 1637069
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1986303

 
alfa aesar™
L1986303
1g Each for $238.59

AAL1986306

 
alfa aesar™
L1986306
5g Each for $938.96
Description & Specifications

Specifications

Formula Weight 158.54
Molecular Formula C5H3ClN2O2
MDL Number MFCD00490110
CAS 54231-32-2
Beilstein 1637069
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Chemical Name or Material 3-Chloro-2-nitropyridine
Assay 97%
Melting Point 89° to 92°C
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.