Please login to your online account to display your discounted pricing

3-Chloro-2-nitrobenzoic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-nitrobenzoic acid
CAS 4771-47-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H4ClNO4
Linear Formula ClC6H3(NO2)CO2H
Formula Weight 201.57
MDL Number MFCD00007087
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC151690050

  SDS
thermo fisher scientific
151690050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC151690010

  SDS
thermo fisher scientific
AC151690010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 201.57
Quantity 5g
Color Almost white to beige
Melting Point 236° to 240°C
CAS Max % 100.0
Chemical Name or Material 3-Chloro-2-nitrobenzoic acid
Assay Percent Range 98%
Linear Formula ClC6H3(NO2)CO2H
MDL Number MFCD00007087
CAS 4771-47-5
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C7H4ClNO4
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.