missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-2-methylpropene (Tech.), 90%, ACROS Organics™

Manufacturer:  Acros Organics 148420500

 View more versions of this product

Catalog No. AC148420500


 

Specifications

Specifications

3-Chloro-2-methylpropene
563-47-3
100.0
Glass bottle
0.1% max.
≥88%
90%
C4H7Cl
MFCD00000953
15, 2150
OHXAOPZTJOUYKM-UHFFFAOYSA-N
3-chloro-2-methylprop-1-ene
11241
90.55
tech., 90%
88.0
Authentic
1.4262 to 1.4282 (20°C, 589nm)
Technical Grade
Undesignated
50mL
H2C=C(CH3)CH2Cl
01, 209
Methallyl chloride
CC(=C)CCl
90.55
CHEBI:82419
Liquid

Chemical Identifiers

563-47-3
90.55
OHXAOPZTJOUYKM-UHFFFAOYSA-N
11241
C4H7Cl
MFCD00000953
Methallyl chloride
CHEBI:82419
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. May cause an allergic skin reaction. Harmful if swallowed. Harmful if inhaled. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapour.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger

EINECSNumber : 209-251-2

RTECSNumber : UC8050000

TSCA : TSCA

SDS Documents