Please login to your online account to display your discounted pricing

3-Chloro-2-methylpropene (Tech.), 90%, ACROS Organics™

$30.41 - $87.26

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-methylpropene
Name Note tech., 90%
CAS 563-47-3
CAS Min % 88.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥88%
Assay Percent Range 90%
Molecular Formula C4H7Cl
Linear Formula H2C=C(CH3)CH2Cl
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC148420010

  SDS
thermo fisher scientific
148420010
Glass bottle 1L Each for $87.26

AC148420500

  SDS
thermo fisher scientific
148420500
Glass bottle 50mL Each for $31.04

AC148422500

  SDS
thermo fisher scientific
148422500
Glass bottle 250mL Each for $30.41
Description & Specifications

Specifications

Purity Grade Technical Grade
Physical Form Liquid
Assay ≥88%
Formula Weight 90.55
Quantity 1L
Color Clear
Beilstein 01, 209
Merck Index 15, 2150
EINECS Number 209-251-2
TSCA TSCA
RTECS Number UC8050000
CAS Min % 88.0
CAS Max % 100.0
Water 0.1% max.
Assay Percent Range 90%
Linear Formula H2C=C(CH3)CH2Cl
Health Hazard 1 Danger
Infrared Spectrum Authentic
Chemical Name or Material 3-Chloro-2-methylpropene
Refractive Index 1.4262 to 1.4282 (20°C, 589nm)
Name Note tech., 90%
Molecular Formula C4H7Cl
MDL Number MFCD00000953
CAS 563-47-3
Synonym Methallyl chloride
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May cause an allergic skin reaction. Harmful if swallowed. Harmful if inhaled. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapo
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact
Packaging Glass bottle
Safety and Handling

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact

GHS P Statement Causes severe skin burns and eye damage. May cause an allergic skin reaction. Harmful if swallowed. Harmful if inhaled. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapo