Please login to your online account to display your discounted pricing

3-chloro-2-methylphenyl isothiocyanate, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-methylphenyl isothiocyanate
CAS 19241-35-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6ClNS
Formula Weight 183.66
MDL Number MFCD00022056
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00697EA

 
fisher scientific company
TL00697EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.66
Quantity 10g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
CAS 19241-35-1
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Chemical Name or Material 3-Chloro-2-methylphenyl isothiocyanate
MDL Number MFCD00022056
Molecular Formula C8H6ClNS