missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-methylbenzonitrile, 97%

Manufacturer:  Alfa Aesar™ L1104803

 View more versions of this product

Catalog No. AAL1104803



Specifications

Specifications

3-Chloro-2-methylbenzonitrile
20°C to 23°C
54454-12-5
MFCD00045598
2613370
FKFZTNLSUJCIMG-UHFFFAOYSA-N
3-chloro-2-methylbenzonitrile
521508
97%
120°C (22mmHg)
1g
C8H6ClN
UN3439
2-methyl-3-chlorobenzonitrile, pubchem23847, 3-chloro-o-tolunitrile, acmc-1aqtx, 6-chloro-2-cyanotoluene, 2-chloro-6-cyanotoluene, 2-methyl-3-chloro-benzonitrile, 3-chloro-2-methyl-benzonitrile, benzonitrile,3-chloro-2-methyl, benzonitrile, 3-chloro-2-methyl
CC1=C(C=CC=C1Cl)C#N
151.593
151.6

Chemical Identifiers

54454-12-5
151.593
FKFZTNLSUJCIMG-UHFFFAOYSA-N
521508
CC1=C(C=CC=C1Cl)C#N
C8H6ClN
MFCD00045598
2-methyl-3-chlorobenzonitrile, pubchem23847, 3-chloro-o-tolunitrile, acmc-1aqtx, 6-chloro-2-cyanotoluene, 2-chloro-6-cyanotoluene, 2-methyl-3-chloro-benzonitrile, 3-chloro-2-methyl-benzonitrile, benzonitrile,3-chloro-2-methyl, benzonitrile, 3-chloro-2-methyl
3-chloro-2-methylbenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents