Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-methylbenzoic acid, 98%

$185.34 - $185.34

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-methylbenzoic acid
CAS 7499-08-3
Molecular Formula C8H7ClO2
Formula Weight 170.6
MDL Number MFCD00053312
Beilstein 1072826
Synonym 3-Chloro-o-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1577306

  SDS
alfa aesar™
L1577306
5g Each for $185.34
Description & Specifications

Specifications

Formula Weight 170.6
Chemical Name or Material 3-Chloro-2-methylbenzoic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 3-Chloro-o-toluic acid
MDL Number MFCD00053312
Molecular Formula C8H7ClO2
Beilstein 1072826
RTECS Number XU1645000
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Quantity 5g
Melting Point 162° to 166°C
CAS 7499-08-3
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.