Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-methylbenzenesulfonyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-methylbenzenesulfonyl chloride
CAS 80563-86-6
Molecular Formula C7H6Cl2O2S
Formula Weight 225.09
MDL Number MFCD00052198
UN Number UN3261
Beilstein 3261529
Synonym 3-Chloro-o-toluenesulfonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1332203

 
alfa aesar™
A1332203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1332206

 
alfa aesar™
A1332206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1332214

 
alfa aesar™
A1332214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.09
Chemical Name or Material 3-Chloro-2-methylbenzenesulfonyl chloride
Assay 98%
Molecular Formula C7H6Cl2O2S
MDL Number MFCD00052198
CAS 80563-86-6
UN Number UN3261
Beilstein 3261529
Synonym 3-Chloro-o-toluenesulfonyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Melting Point 69° to 73°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.