Please login to your online account to display your discounted pricing

3-Chloro-2-methylbenzenesulfonyl chloride, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-methylbenzene-1-sulfonyl chloride
Assay 95%
Assay Percent Range 95%
Molecular Formula C7H6Cl2O2S
Linear Formula Cl(CH3)C6H3SO2Cl
Formula Weight 225.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00453EA

 
fisher scientific company
TL00453EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.09
Quantity 10g
Assay 95%
Chemical Name or Material 3-Chloro-2-methylbenzene-1-sulfonyl chloride
Assay Percent Range 95%
Molecular Formula C7H6Cl2O2S
Linear Formula Cl(CH3)C6H3SO2Cl
CAS 80563-86-6