Learn More
3-Chloro-2-methylanisole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2220314
Description
Arene-manganese tricarbonyl complexes are prepared using 3-Chloro-2-methylanisole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsArene-manganese tricarbonyl complexes are prepared using 3-Chloro-2-methylanisole.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
3-Chloro-2-methylanisole | |
-3°C | |
212°C to 215°C | |
C8H9ClO | |
MFCD00070772 | |
UN1993 | |
3-chloro-2-methylanisole, 2-methyl-3-chloroanisole, 2-chloro-6-methoxytoluene, benzene, 1-chloro-3-methoxy-2-methyl, 2-methoxy-6-chlorotoluene, 1-chloro-3-methoxy-2-methyl-benzene, pubchem2664, acmc-1cq3o, ltvrgawoeokgjz-uhfffaoysa | |
COC1=CC=CC(Cl)=C1C | |
156.61 | |
156.61 |
3260-88-6 | |
1.16 | |
92°C (197°F) | |
1.5395 | |
25 g | |
1862220 | |
LTVRGAWOEOKGJZ-UHFFFAOYSA-N | |
1-chloro-3-methoxy-2-methylbenzene | |
76749 | |
97% |
Chemical Identifiers
3260-88-6 | |
156.61 | |
LTVRGAWOEOKGJZ-UHFFFAOYSA-N | |
76749 | |
COC1=CC=CC(Cl)=C1C |
C8H9ClO | |
MFCD00070772 | |
3-chloro-2-methylanisole, 2-methyl-3-chloroanisole, 2-chloro-6-methoxytoluene, benzene, 1-chloro-3-methoxy-2-methyl, 2-methoxy-6-chlorotoluene, 1-chloro-3-methoxy-2-methyl-benzene, pubchem2664, acmc-1cq3o, ltvrgawoeokgjz-uhfffaoysa | |
1-chloro-3-methoxy-2-methylbenzene |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 221-862-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only