missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-2-methylaniline, TCI America™

$25.97 - $154.31

Chemical Identifiers

CAS 87-60-5
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00007766
InChI Key ZUVPLKVDZNDZCM-UHFFFAOYSA-N
Synonym 3-chloro-o-toluidine, 2-amino-6-chlorotoluene, benzenamine, 3-chloro-2-methyl, 3-chloro-2-toluidine, scarlet tr base, fast scarlet tr base, 6-chloro-2-aminotoluene, o-toluidine, 3-chloro, 3-chloro-2-methylphenylamine, o-methyl-m-chloroaniline
PubChem CID 6894
IUPAC Name 3-chloro-2-methylaniline
SMILES CC1=C(C=CC=C1Cl)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C030025ML View Documents TCI America
C030025ML
25mL Each for $25.97
 
C0300500ML View Documents TCI America
C0300500ML
500mL Each for $154.31
 
Specifications

Chemical Identifiers

87-60-5
141.598
ZUVPLKVDZNDZCM-UHFFFAOYSA-N
6894
CC1=C(C=CC=C1Cl)N
C7H8ClN
MFCD00007766
3-chloro-o-toluidine, 2-amino-6-chlorotoluene, benzenamine, 3-chloro-2-methyl, 3-chloro-2-toluidine, scarlet tr base, fast scarlet tr base, 6-chloro-2-aminotoluene, o-toluidine, 3-chloro, 3-chloro-2-methylphenylamine, o-methyl-m-chloroaniline
3-chloro-2-methylaniline

Specifications

245°C
3°C
87-60-5
MFCD00007766
3-chloro-o-toluidine, 2-amino-6-chlorotoluene, benzenamine, 3-chloro-2-methyl, 3-chloro-2-toluidine, scarlet tr base, fast scarlet tr base, 6-chloro-2-aminotoluene, o-toluidine, 3-chloro, 3-chloro-2-methylphenylamine, o-methyl-m-chloroaniline
CC1=C(C=CC=C1Cl)N
141.598
141.60
≥99.0% (GC,T)
Brown
25mL
C7H8ClN
3429
ZUVPLKVDZNDZCM-UHFFFAOYSA-N
3-chloro-2-methylaniline
6894
Clear Liquid at 20°C
3-Chloro-2-methylaniline
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0265

RTECSNumber : XU4760000

TSCA : Yes