Please login to your online account to display your discounted pricing

3-Chloro-2-methylaniline 99%, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC146490025

 View more versions of this product

Catalog No. AC146490025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.5%
Formula Weight 141.6
Quantity 2.5L
Color Clear yellow to red or red-brown
EINECS Number 201-756-6
TSCA TSCA
RTECS Number XU4760000
Density 1.171g/mL
Beilstein 12, 836
CAS Max % 100.0
Storage Information Product may darken during storage
Infrared Spectrum Authentic
Synonym 3-Chloro-o-toluidine
Packaging Glass bottle
Chemical Name or Material 3-Chloro-2-methylaniline
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H8ClN
Linear Formula ClC6H3(CH3)NH2
MDL Number MFCD00007766
CAS 87-60-5
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Refractive Index 1.5870 to 1.5890 (20°C, 589nm)

Specifications

Chemical Name or Material 3-Chloro-2-methylaniline
Name Note 99%
CAS 87-60-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H8ClN
Linear Formula ClC6H3(CH3)NH2
Formula Weight 141.6
MDL Number MFCD00007766
Synonym 3-Chloro-o-toluidine
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.

SDS & Safety Documents