missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Chloro-2-methoxybenzoic Acid 98.0+%, TCI America™
$187.46 - $572.35
Chemical Identifiers
CAS | 3260-93-3 |
---|---|
Molecular Formula | C8H7ClO3 |
Molecular Weight (g/mol) | 186.59 |
MDL Number | MFCD00179306 |
InChI Key | NKVUYEGKHRDEBB-UHFFFAOYSA-N |
Synonym | 3-Chloro-o-anisic Acid |
PubChem CID | 6423044 |
IUPAC Name | 3-chloro-2-methoxybenzoic acid |
SMILES | COC1=C(C=CC=C1Cl)C(O)=O |
Chemical Identifiers
3260-93-3 | |
186.59 | |
NKVUYEGKHRDEBB-UHFFFAOYSA-N | |
6423044 | |
COC1=C(C=CC=C1Cl)C(O)=O |
C8H7ClO3 | |
MFCD00179306 | |
3-Chloro-o-anisic Acid | |
3-chloro-2-methoxybenzoic acid |
Specifications
3260-93-3 | |
White-Yellow | |
MFCD00179306 | |
3-Chloro-o-anisic Acid | |
COC1=C(C=CC=C1Cl)C(O)=O | |
186.59 | |
186.59 | |
Crystalline Powder |
122°C | |
C8H7ClO3 | |
1 g | |
NKVUYEGKHRDEBB-UHFFFAOYSA-N | |
3-chloro-2-methoxybenzoic acid | |
6423044 | |
≥98.0% (GC,T) | |
3-Chloro-2-methoxybenzoic Acid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator