Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-iodotoluene, 98%

$61.64 - $212.46

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-iodotoluene
CAS 5100-98-1
Molecular Formula C7H6ClI
Formula Weight 252.48
MDL Number MFCD00051756
Beilstein 2961541
Synonym 1-Chloro-2-iodo-3-methylbenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0682403

  SDS
alfa aesar™
L0682403
1g Each for $61.64

AAL0682406

  SDS
alfa aesar™
L0682406
5g Each for $212.46
Description & Specifications

Specifications

Formula Weight 252.48
Chemical Name or Material 3-Chloro-2-iodotoluene
Assay 98%
Molecular Formula C7H6ClI
MDL Number MFCD00051756
CAS 5100-98-1
Synonym 1-Chloro-2-iodo-3-methylbenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Melting Point 25° to 29°C
Quantity 1g
Beilstein 2961541
Boiling Point 134° to 136°C (20mmHg)
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.