Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-hydroxypyridine, 97%

$67.81 - $849.81

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-hydroxypyridine
CAS 13466-35-8
Molecular Formula C5H4ClNO
Formula Weight 129.55
MDL Number MFCD01646109
Beilstein 109845
Synonym 3-Chloro-2-pyridinol; 3-Chloro-2(1H)-pyridone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2002703

  SDS
alfa aesar™
L2002703
1g Each for $67.81

AAL2002706

  SDS
alfa aesar™
L2002706
5g Each for $224.51

AAL2002714

  SDS
alfa aesar™
L2002714
25g Each for $849.81
Description & Specifications

Specifications

Formula Weight 129.55
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Synonym 3-Chloro-2-pyridinol; 3-Chloro-2(1H)-pyridone
Molecular Formula C5H4ClNO
Chemical Name or Material 3-Chloro-2-hydroxypyridine
CAS 13466-35-8
Beilstein 109845
Quantity 1g
MDL Number MFCD01646109
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 180° to 182°C
Assay 97%

Applications
3-Chloro-2-hydroxypyridine is used as an organic intermediate.

Solubility
Soluble in water (25 deg C)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.