Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluoropyridine-4-boronic acid pinacol ester, 95%

$420.58 - $420.58

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluoropyridine-4-boronic acid pinacol ester
CAS 1073353-71-5
Molecular Formula C11H14BClFNO2
Formula Weight 257.50g/mol
MDL Number MFCD09037476
Synonym 3-Chloro-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2745703

 
alfa aesar™
H27457-03
1g Each for $420.58
Description & Specifications

Specifications

Melting Point 76° to 80°C
Quantity 1g
Chemical Name or Material 3-Chloro-2-fluoropyridine-4-boronic acid pinacol ester
CAS 1073353-71-5
Assay 95%
Molecular Formula C11H14BClFNO2
Formula Weight 257.50g/mol
MDL Number MFCD09037476
Synonym 3-Chloro-2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.