Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluoropyridine-4-boronic acid, 95%

$510.80 - $510.80

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluoropyridine-4-boronic acid
CAS 1217500-55-4
Molecular Formula C5H4BClFNO2
Formula Weight 175.35
MDL Number MFCD09037477
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2778103

  SDS
alfa aesar™
H2778103
1g Each for $510.80
Description & Specifications

Specifications

Formula Weight 175.35
Quantity 1g
Molecular Formula C5H4BClFNO2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 126° to 130°C
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 1217500-55-4
Health Hazard 1 Warning
Chemical Name or Material 3-Chloro-2-fluoropyridine-4-boronic acid
Assay 95%
MDL Number MFCD09037477
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.