Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluorophenylacetic acid, 97%

$183.82 - $183.82

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluorophenylacetic acid
CAS 261762-96-3
Molecular Formula C8H6ClFO2
Formula Weight 188.59
MDL Number MFCD01631554
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2646503

  SDS
alfa aesar™
H2646503
1g Each for $183.82
Description & Specifications

Specifications

Formula Weight 188.59
Chemical Name or Material 3-Chloro-2-fluorophenylacetic acid
Assay 97%
Molecular Formula C8H6ClFO2
MDL Number MFCD01631554
CAS 261762-96-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.