Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluorobenzyl bromide, 96%

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluorobenzyl bromide
CAS 85070-47-9
Molecular Formula C7H5BrClF
Formula Weight 223.48
MDL Number MFCD00042503
UN Number UN3261
Synonym alpha-Bromo-3-chloro-2-fluorotoluene; alpha-Bromo-2-fluoro-3-chlorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1311403

  SDS
alfa aesar™
A1311403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1311406

  SDS
alfa aesar™
A1311406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 223.48
Assay 96%
Boiling Point 125° to 126°C (15mmHg)
Quantity 1g
MDL Number MFCD00042503
Synonym alpha-Bromo-3-chloro-2-fluorotoluene; alpha-Bromo-2-fluoro-3-chlorotoluene
Molecular Formula C7H5BrClF
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Melting Point 36° to 40°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Health Hazard 1 Danger
CAS 85070-47-9
Chemical Name or Material 3-Chloro-2-fluorobenzyl bromide
UN Number UN3261
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.