Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluorobenzoic acid, 97%

$150.68 - $497.23

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluorobenzoic acid
CAS 161957-55-7
Molecular Formula C7H4ClFO2
Formula Weight 174.56
MDL Number MFCD00042506
Beilstein 7127637
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2060303

  SDS
alfa aesar™
B2060303
1g Each for $150.68

AAB2060306

  SDS
alfa aesar™
B2060306
5g Each for $497.23
Description & Specifications

Specifications

Formula Weight 174.56
Chemical Name or Material 3-Chloro-2-fluorobenzoic acid
Assay 97%
Molecular Formula C7H4ClFO2
MDL Number MFCD00042506
CAS 161957-55-7
Beilstein 7127637
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 176° to 178°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.