Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluorobenzenesulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluorobenzenesulfonyl chloride
CAS 351003-48-0
Molecular Formula C6H3Cl2FO2S
Formula Weight 229.06
MDL Number MFCD03094234
UN Number UN3265
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1951103

 
alfa aesar™
L1951103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1951106

 
alfa aesar™
L1951106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.06
MDL Number MFCD03094234
CAS 351003-48-0
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point >110°C (230°F)
Chemical Name or Material 3-Chloro-2-fluorobenzenesulfonyl chloride
Assay 97%
Molecular Formula C6H3Cl2FO2S
UN Number UN3265
Health Hazard 1 Danger
Quantity 1g
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store in dry condition. Keep oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.