Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluorobenzamide, 97+%

$338.64 - $338.64

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluorobenzamide
CAS 104326-94-5
Molecular Formula C7H5ClFNO
Formula Weight 173.58
MDL Number MFCD00042205
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2416209

 
alfa aesar™
B2416209
10g Each for $338.64
Description & Specifications

Specifications

Formula Weight 173.58
Chemical Name or Material 3-Chloro-2-fluorobenzamide
Assay ≥97%
Molecular Formula C7H5ClFNO
MDL Number MFCD00042205
CAS 104326-94-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 124° to 127°C
Quantity 10g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.