Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluorobenzaldehyde, 97%

$65.41 - $751.88

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluorobenzaldehyde
CAS 85070-48-0
Molecular Formula C7H4ClFO
Formula Weight 158.56
MDL Number MFCD01631571
Beilstein 5861249
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAB2297503

  SDS
alfa aesar™
B2297503
Each for $65.41

AAB2297506

  SDS
alfa aesar™
B2297506
Each for $198.89

AAB2297514

  SDS
alfa aesar™
B2297514
Each for $751.88
Description & Specifications

Specifications

Formula Weight 158.56
Chemical Name or Material 3-Chloro-2-fluorobenzaldehyde
Assay 97%
Molecular Formula C7H4ClFO
MDL Number MFCD01631571
CAS 85070-48-0
Beilstein 5861249
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.