Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluoroanisole, 97%

$129.27 - $390.26

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluoroanisole
CAS 261762-56-5
Molecular Formula C7H6ClFO
Formula Weight 160.58
MDL Number MFCD01631570
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2656003

  SDS
alfa aesar™
H2656003
1g Each for $129.27

AAH2656006

  SDS
alfa aesar™
H2656006
5g Each for $390.26
Description & Specifications

Specifications

Formula Weight 160.58
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 261762-56-5
Assay 97%
MDL Number MFCD01631570
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C7H6ClFO
Quantity 1g
Chemical Name or Material 3-Chloro-2-fluoroanisole
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.